Learn More
Air Science Usa Base Stand 24" Wide for Horizontal Laminar Flow Cabinet
SDP
Supplier:  Air Science Usa CARTBWHLF24
Encompass_Preferred
Base Stand, (Metal), Open Frame, Adjustable Leveling Feet, Fixed height. Fits Hlf-24 / Hlf-24XT Models
Catalog No. NC2684341