Learn More
Air Science Usa Base Stand 48" Wide for Horizontal Laminar Flow Cabinet
SDP
Supplier:  Air Science Usa CARTBWHLF48
Encompass_Preferred
Base Stand, (Metal), Open Frame, Adjustable Leveling Feet, Fixed height. Fits LF HLF-48 / HLF-48XT Models
Catalog No. NC2684345