Learn More
Air Science Usa Base Stand 60" Wide for Horizontal Laminar Flow Cabinet
SDP
Supplier:  Air Science Usa CARTBWHLF60
Encompass_Preferred
Base Stand, (Metal), Open Frame, Adjustable Leveling Feet, Fixed height. Fits Lf Hlf-60 / Hlf-60XT Models
Catalog No. NC2684346