Learn More
Wilmad Labglass Support Base, Fritted Glass. Filter Size: 47mm
Supplier:  Wilmad Labglass BP1752001
Encompass_Preferred
Support Base, Fritted Glass. Filter Size: 47mm. Replacement membrane filter Support base for BP-1755-000 Microfiltration Glassware. Supplied with coarse porosity glass frit.
Catalog No. 50-804-637
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov