Learn More
Ace Glass, Inc. BASE SYSTEM,100L LP JKTD "C"
SDP
Supplier:  Ace Glass, Inc. 1284517
Encompass
Jacketed reactor base system designed for maximum diversity and ease of use. Ace Glass has developed a simple base system building platform which allows any reactor system to be customized using catalog and/or custom designed parts. Each base system comes with the basic starting components required. Simply select the motor and accessories needed to complete the design. For customized components or application design, contact our technical department for further assistance.
Catalog No. 50-008-54796