Learn More
Selleck Chemical LLC Basimglurant E4619-100MG
SDP
Supplier:  Selleck Chemical LLC E4619100MG
Encompass_Preferred
Basimglurant(RG7090 CTEP Derivative) is a potent and selective negative allosteric modulator of mGlu5 with a Kd of 1 1 nM It inhibits the function of mGluR5 thereby regulating glutamatergic neurotransmission
Catalog No. 50-000-39410