Learn More
Cayman Chemical BasInolIde 2.5mg
SDP
Supplier:  Cayman Chemical 220012.5 MG
Encompass_Preferred
A cyclodepsipeptide insecticide; synthesized by B. bassiana and V. lecanii; induces atony in B. mori when given orally at concentrations as low as 4 ppm; inhibits the muscarinic, but not nicotinic, receptor-induced contractions in response to acetylcholine in smooth muscle
Catalog No. 50-296-4326