Learn More
Bioworld Basta Solution, 1%, 100 mL
Supplier:  Bioworld 306320061
Encompass_Preferred
BastaSynonym: glufosinateNon-selective herbicide for plant transformation studies using nucleotide sequence of the phosphinothricin N-acetyltransferase gene from Streptomyces viridochromogenes, "Bar gene"Acts by inhibiting glutamine synthetase and thus blocks all nitrogen assimilation into the plantAppearance: clear liquid.Melting Point: 209-215 °CSoluble in waterBasta is the registered trademark of Bayer CropScience
Catalog No. NC9061701