Learn More
Hach Company Battery Charger for TSS Portable Handheld Instrument
Supplier:  Hach Company LZY607
Encompass
The Hach LZY607 battery charger charges the rechargeable batteries installed in the TSS Portable handheld turbidity/TSS instrument. The adapter includes plug inserts for four different power plug regions, enabling charging in multiple countries.
  • Charges rechargeable batteries while they remain installed in the portable instrument.
  • Includes plug inserts for four power plug regions for international compatibility.
  • Provides a simple adapter connection for convenient field charging.
Catalog No. NC2279130