Learn More
Lascar Electronics Inc Replacement Battery for EL-USB-1-PRO
Supplier: Lascar Electronics Inc BAT 3V6 1/2AA
Non-rechargeable 1/2 AA lithium battery for use with most Lascar data loggers (excluding EL-USB-1-PRO, EL-USB-1-RCG, EL-USB-LITE & EL-USB-RT)