Learn More
Essendant BATTERY2025 LITH100SV
Supplier:  Essendant EVEECRN2025
Encompass
Energizer Industrial 2025 Lithium Batteries deliver the power your specialty professional devices need Construction site facility maintenance medical center business office and classroom wherever the job takes you trust your precision electronics to lithium 2025 batteries that are 100pct leakproof in standard usage They operate in worksite temperatures ranging from -40 F to 140 F These coin batteries hold their power with up to 10 years of shelf life so you won t have device downtime When work depends on calculators digital thermometers glucose monitors and more you can trust the power and protection of Energizer Industrial 2025 Lithium Batteries Battery Type- Lithium Battery Size- CR2025 For Use With- Car Remotes Digital Thermometers Digital Watches Glucose Monitors Battery Capacity- 165 mAh
Catalog No. 50-000-35599