Learn More
Essendant BATTERYAARECHG2
Supplier:  Essendant DURNLAA2BCD
Encompass_Preferred
Duracell Rechargeable Pre-Charged NiMH batteries let you charge hundreds of times helping you save money With its Ion Core technology you get amazing power in every battery Plus they come pre-charged and ready to use They are guaranteed to last for 10 years (or up to 400 recharges whichever comes first) in storage and they will hold a charge for up to one year when not in use These powerful NiMH batteries work in any NiMH charger and you can recharge them up to 400 times Battery Type- NiMH Rechargeable Battery Size- AA For Use With- Everyday Electronic Devices Battery Capacity- 2400 mAh
Catalog No. 50-000-18660