Learn More
Essendant BATTERYELN123LITH12SV
Supplier:  Essendant EVEELN12312
Encompass_Preferred
Power your specialty professional devices with Energizer Industrial Lithium 123 Batteries Construction site facility maintenance medical center business office and classroom wherever the job takes you trust your precision electronics to Lithium 123 batteries that are 100pct leakproof in standard usage They operate in worksite temperatures ranging from -40 F to 140 F They hold their power with up to 10 years of shelf life so you won t have device downtime When work depends on motion sensors security cameras two-way radios and more you can trust the power and protection of Energizer Industrial Lithium 123 Batteries Battery Type- Lithium Battery Size- CR123 For Use With- Cameras Flashlights Motion Sensors Security Cameras Small Electronics Two-way Radios Battery Capacity- 1500 mAh
Catalog No. 50-000-35739