Learn More
Essendant BATTERYELN1CR28LITH8SV
Supplier:  Essendant EVEELN1CR28
Encompass
Rely on Energizer Industrial Lithium CR2 Batteries to power your specialty professional devices Construction site facility maintenance medical center business office and classroom wherever the job takes you trust your precision electronics to lithium CR2 batteries that are 100pct leakproof in normal usage They operate in work site temperatures ranging from -40 F to 140 F They hold their power with up to 10 years of shelf life so you won t have device downtime When work depends on laser pointers motion sensors automatic paper towel dispensers and more you can rely on the power and protection of Energizer Industrial Lithium CR2 Batteries Battery Type- Lithium Battery Size- CR2 For Use With- Automatic Paper Towel Dispensers Cameras Laser Pointers Motion Sensors Photography Equipment Battery Capacity- 750 mAh
Catalog No. 50-000-35740