Learn More
Essendant BATTERYLN91LITHAA4SV
Supplier:  Essendant EVELN91PK
Encompass
Power your industrial and professional devices with Energizer Industrial Lithium AA Batteries Construction site facility maintenance medical center business office and classroom-wherever you work the longest-lasting professional AA batteries keep your electronics up for the job In normal use they re 100pct leakproof to protect your worksite devices and they operate in temperatures ranging from -40 F to 140 F With a 25-year shelf life you won t have device downtime All of that lithium performance comes in a heavy-duty AA battery that weighs 33pct less than an alkaline AA When work depends on automatic paper towel dispensers hard hat lights pulse oximeters and more you can rely on the power and protection of Energizer Industrial Lithium AA Batteries Battery Type- Lithium Battery Size- AA For Use With- Automatic Paper Towel Dispensers Flashlights Hard Hat Lights Pulse Oximeters Remotes Small Electrical Equipment Battery Capacity- 3500 mAh
Catalog No. 50-000-35733