Learn More
Apexbio Technology LLC Batyl alcohol 544-62-7 100mg
SDP
Supplier:  Apexbio Technology LLC C6759100
Encompass_Preferred
Batyl alcohol (CAS 544-62-7) is an endogenous alkylglycerol compound present in various biological tissues It functions as a metabolic intermediate in the biosynthesis of phospholipids and ether lipids thereby influencing membrane fluidity and cellular signaling pathways In biomedical research batyl alcohol is frequently employed to study lipid metabolic processes the structural and functional roles of biomembranes and cellular events such as proliferation differentiation and apoptosis Additionally it serves as a molecular probe to investigate inflammatory responses immune regulation and lipid metabolism disorders
Catalog No. 50-000-90134