Learn More
Selleck Chemical LLC Batyl alcohol S3982-25mg
SDP
Supplier:  Selleck Chemical LLC S398225MG
Encompass_Preferred
Batyl alcohol (Batilol Stearyl monoglyceride 1-O-Octadecylglycerol Batilolum) isolated from shark liver oil is also present in yellow bone marrow of animals and has been described as an inflammatory agent
Catalog No. 50-310-8600