Learn More
Selleck Chemical LLC Bavdegalutamide (ARV-110) | 98.78% | 2222112-77-6 | 25mg
SDP
Supplier:  Selleck Chemical LLC S696525MG
Encompass_Preferred
Bavdegalutamide (ARV-110), an orally bioavailable, specific androgen receptor (AR) PROTAC degrader that leads to ubiquitination and degradation of AR. ARV-110 completely degrades androgen receptor (AR) in all cell lines tested with DC50 of < 1 nM. ARV-110 can be used for the research of prostate cancer.
Catalog No. NC2224503