Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bax protein, human (His) | >95.0% | 100 UG
SDP

Supplier:  Medchemexpress LLC HYP7651100UG

Encompass_Preferred

Recombinant human Bax protein (His-tag) is a full-length apoptosis regulator (BAX, UniProt Q07812) produced in E. coli and supplied as a stabilized solution for biochemical and cell biology research. It corresponds to amino acids M1-Q171 and has an approximate molecular weight of 22.0 kDa.

  • His-tagged full-length human BAX (M1-Q171).
  • Expressed in E. coli and supplied as a filtered solution in 20 mM PB, 150 mM NaCl, 2 mM EDTA, 5% trehalose, 30% glycerol, pH 7.4.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 22.0 kDa.
  • Store at -80 °C for long term; avoid repeated freeze-thaw cycles; ship on dry ice.
  • Available in 100 μg format for experimental use.

Catalog No. 50-002-90332


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.