Learn More
Selleck Chemical LLC BAY 1217389 5mg 1554458-53-5
SDP
Supplier:  Selleck Chemical LLC S82155MG
Encompass_Preferred
BAY 1217389 is an orally bioavailable, selective inhibitor of the serine/threonine kinase monopolar spindle 1 (Mps1) with IC50 values below 10 nmol/L while showing an excellent selectivity profile. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5808