Learn More
Medchemexpress LLC HY-100020 50mg Medchemexpress, BAY-1436032 CAS:1803274-65-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10002050MG
Encompass_Preferred
Medchemexpress, HY-100020 50mg BAY-1436032 CAS:1803274-65-8 BAY-1436032 is a novel pan-mutant isocitrate dehydrogenase 1 ( IDH1 ) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1694