Learn More
Medchemexpress LLC HY-112645 5mg Medchemexpress, BAY-2402234 CAS:2225819-06-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1126455MG
Encompass_Preferred
Medchemexpress, HY-112645 5mg BAY-2402234 CAS:2225819-06-5 BAY-2402234 is a selective dihydroorotate dehydrogenase (DHODH) inhibitor for the treatment of myeloid malignancies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9129