Learn More
Selleck Chemical LLC BAY-293 S8826-100MG
SDP
Supplier:  Selleck Chemical LLC S8826100MG
Encompass_Preferred
BAY-293 selectively inhibits the KRAS-SOS1 interaction with an IC50 of 21 nM
Catalog No. 50-000-24740