Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bay-298 | 2471978-97-7 | 100.0% | 473.93 g/mol | C27H21ClFN3O2 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1302495MG

Encompass_Preferred

BAY-298 is an orally active, selective luteinizing hormone receptor (LH-R) antagonist used as a research compound to probe LH-R biology and to reduce sex hormone levels in vitro and in vivo. It is supplied as a high-purity solid for laboratory research use only.

  • Selective luteinizing hormone receptor antagonist.
  • Orally active compound with demonstrated in vivo effects.
  • Reported IC50s: hLH 96 nM, rLH 23 nM, cLH 78 nM.
  • High purity (reported 99.96%).
  • Molecular formula C27H21ClFN3O2; molecular weight 473.93 g/mol.
  • Supplied in small milligram pack sizes suitable for preclinical studies.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-63900


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.