Learn More
Medchemexpress LLC BAY-298 | 2471978-97-7 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13024950MG
Encompass_Preferred
BAY-298 is an orally active and selective antagonist of the luteinizing hormone receptor (LH-R). It exhibits IC50 values of 96 nM for human LH (hLH), 23 nM for rat LH (rLH), and 78 nM for cynomolgus monkey LH (cLH). This compound has demonstrated the ability to reduce sex hormone levels and is intended for research use only.
  • Orally active and selective LH-R antagonist
  • Inhibits human, rat, and cynomolgus monkey LH-R
  • Reduces sex hormone levels
  • For research use only
Catalog No. 50-003-39894