Learn More
Medchemexpress LLC Bay-320 | 1445830-50-1 | MFCD31657346 | 99.7% | 492.52 g·mol⁻1 | C26H26F2N6O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10400010MM 1ML
Encompass_Preferred
BAY-320 is a selective, ATP-competitive inhibitor of the Bub1 kinase used in cellular and cancer research. It inhibits Bub1 kinase activity (reported IC50 680 nM), disrupts Sgo1 localization during mitosis, and affects chromosome alignment and spindle assembly. The compound is supplied as a 10 mM solution in DMSO and is provided with analytical data and storage recommendations.
  • Selective ATP-competitive Bub1 kinase inhibitor with reported IC50 of 680 nM.
  • Disrupts Sgo1 localization and mitotic chromosome alignment.
  • Suitable for cellular studies related to mitosis and cancer research.
  • Provided as a 10 mM solution in DMSO for ready-to-use applications.
  • High reported purity with supporting analytical data available.
  • Stable when stored according to supplier recommendations.
Catalog No. 50-002-44145