Learn More
Cayman Chemical BAY 412272 Nitric Oxide Reagen
SDP
Supplier:  Cayman Chemical 1005710 MG
Encompass_Preferred
An activator of sGC, stimulating activity at concentrations as low as 10-100 nM; inhibits platelet aggregation (IC50 = 36 nM), induces relaxation of preconstricted aorta rings (IC50 = 304 nM); effective in vivo, decreasing mean arterial blood pressure in hypertensive rats
Catalog No. 50-288-9646
 Restricted Item