Learn More
AdipoGen BAY 43-9006 (25 mg)
SDP
Supplier:  AdipoGen AGCR10025M025
Encompass_Preferred
Chemical. CAS: 284461-73-0. Formula: C21H16ClF3N4O3. MW: 464.8. Antitumor compound. Broad-spectrum kinase inhibitor. Raf kinase inhibitor. Apoptosis inducer. Cytostatic. Angiogenesis inhibitor. Review.
Catalog No. 50-205-3776