Learn More
Cayman Chemical BAY 57-1293-d4 500UG
SDP
Supplier:  Cayman Chemical 9004311500 UG
Encompass_Preferred
An internal standard for the quantification of BAY 57-1293 by GC- or LC-MS
Catalog No. 50-289-5309