Learn More
Cayman Chemical BAY 59-3074 10mg
SDP
Supplier:  Cayman Chemical 2103610 MG
Encompass_Preferred
A partial agonist of CB1 and CB2 (Kis = 55.4, 48.3, and 45.5 nM for rat CB1, human CB1, and human CB2, respectively); exhibits anti-hyperalgesic and anti-allodynic properties in rat models of chronic neuropathic and inflammatory pain
Catalog No. 50-298-3129