Learn More
TARGETMOL CHEMICALS INC BAY 59-3074 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T369925MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bay 59-3074 is a CB1/CB2 receptor partial agonist (Ki 48.3/45.5 nM). In rat models of chronic neuropathic and inflammatory pain it displays anti-hyperalgesic and antiallodynic properties. purity: 99%
Catalog No. 50-257-6883