Learn More
Medchemexpress LLC Bay 59-3074 | 406205-74-1 | MFCD09027365 | 453.36 | C18H13F6NO4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10048810MM 1ML
Encompass_Preferred
Bay 59-3074 is a selective cannabinoid CB1/CB2 receptor partial agonist supplied as a research reagent for pharmacology studies. The compound exhibits low-nanomolar binding to human CB1 and CB2 receptors and is supplied with formulation, solubility, and storage guidance for both in vitro and in vivo applications.
  • Selective cannabinoid CB1/CB2 partial agonist with low-nanomolar Ki values.
  • Ready-to-use solution format available for immediate assay use.
  • Solid formats available for custom stock preparation and dosing.
  • Solubility data and recommended in vivo formulation guidance included.
  • Documented storage and stability conditions for powder and solutions.
Catalog No. 50-002-14966