Learn More
Selleck Chemical LLC BAY-593 E1976-100MG
SDP
Supplier:  Selleck Chemical LLC E1976100MG
Encompass_Preferred
BAY-593 is an in vivo probe and small molecule inhibitor of geranylgeranyltransferase-I (GGTase-I) blocking oncogenic YAP1/TAZ It exhibits potent anti-tumor activity and inhibits cancer cell proliferation in vitro
Catalog No. 50-000-39148