Learn More
Selleck Chemical LLC Bay 60-7550
SDP
Supplier:  Selleck Chemical LLC S73765MG
Encompass_Preferred
Bay 60-7550 is a potent and selective PDE2 inhibitor with a Ki of 3 8 nM
Catalog No. 50-000-04409