Learn More
Medchemexpress LLC BAY-707 | 2109805-96-9 | 99.6% | 288.34 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112081100MG
Encompass_Preferred
BAY-707 is a chemical probe and a substrate-competitive, highly potent, and selective inhibitor of MTH1 (NUDT1) with an IC50 of 2.3 nM. It exhibits a favorable pharmacokinetic (PK) profile compared to other MTH1 compounds and is well-tolerated in mice, but it lacks in vitro or in vivo anticancer efficacy.
  • Highly potent and selective MTH1 (NUDT1) inhibitor: IC50 of 2.3 nM.
  • Good pharmacokinetic (PK) profile compared to other MTH1 compounds.
  • Well-tolerated in mice with no significant body weight loss observed.
  • Superior cellular target engagement with an EC50 of 7.6 nM.
  • High cell permeability in Caco-2 assay with an efflux ratio of 288 nm/s.
  • Favorable physicochemical profile and promising in vitro pharmacokinetic properties with high metabolic stability in human microsomes and rat hepatocytes.
Catalog No. 50-004-37186