Learn More
Medchemexpress LLC BAY-826 | 1448316-08-2 | 99.9% | 558.53 | C26H19F5N6OS | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100756100MG
Encompass_Preferred
BAY-826 is a selective, potent small-molecule inhibitor of the TIE-2 (TEK) receptor tyrosine kinase developed as a research chemical probe. It exhibits strong binding affinity to TIE-2 (reported Kd ≈ 1.6 nM) and has demonstrated in vitro inhibition of TIE-2 autophosphorylation and in vivo efficacy in preclinical murine glioma models. For research use only; not for human therapeutic use.
  • Potent TIE-2 inhibition (Kd ≈ 1.6 nM).
  • Demonstrated in vivo efficacy in murine glioma models.
  • Suitable for in vitro and in vivo preclinical studies.
  • High chemical purity for pharmacology experiments.
  • Chemical formula C26H19F5N6OS and molecular weight 558.53 g·mol-1.
Catalog No. 50-002-13157