Learn More
Medchemexpress LLC HY-119254 10mg Medchemexpress, BAY-850 CAS:2099142-76-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11925410MG
Encompass_Preferred
Medchemexpress, HY-119254 10mg BAY-850 CAS:2099142-76-2 BAY-850 is a potent and isoform selective ATPase family AAA domain-containing protein 2 (ATAD2) inhibitor, with an IC50 of 166 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9522