Learn More
Selleck Chemical LLC BAY 87-2243 10mg 1227158-85-1
SDP
Supplier:  Selleck Chemical LLC S730910MG
Encompass_Preferred
BAY 87-2243 is a potent and selective hypoxia-inducible factor-1 (HIF-1) inhibitor. BAY 87-2243 inhibits mitochondrial complex I activity, thus triggering a mitophagy-dependent ROS increase leading to necroptosis and ferroptosis. BAY 87-2243 exerts antitumor activity. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4625