Learn More
Selleck Chemical LLC Bay K 8644 5mg
Supplier: Selleck Chemical LLC S79245MG
Bay K 8644 is a potent, selective activator of L-type Ca2+ channel with IC50 of 17.3 nM. *For Research & Development use only. Product is not intended for drug, household, or human consumption.