Learn More
Medchemexpress LLC HY-12859 2mg , BAY1217389 CAS:1554458-53-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY128592MG
Encompass_Preferred
HY-12859 2mg BAY1217389 CAS: 1554458-53-5 BAY 1217389 is a potent, and selective inhibitor of the monopolar spindle 1 (MPS1) kinase with an IC50 value less than 10 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9335