Learn More
Medchemexpress LLC BAY885 | 2307249-33-6 | 99.3% | 515.53 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11208210MM/1ML
Encompass_Preferred
BAY885 is a chemical probe, characterized as a highly potent and selective ERK5 inhibitor with an IC50 of 35 nM. It exhibits weak inhibition on other kinases.
  • Highly potent and selective ERK5 inhibitor.
  • Exhibits an IC50 of 35 nM for ERK5.
  • Shows weak inhibition on other kinases.
  • The ERK5 probe 41 demonstrated potent ERK5 kinase and transcriptional inhibition in the SN12C-MEF2 reporter cell line (IC50 = 115 nM/IC90 = 691 nM).
  • No effects observed on a reporter control cell line with constitutive luciferase expression (SN12C-CMV-luc, IC50 > 30 μM).
Catalog No. 50-004-36368