Learn More
Medchemexpress LLC Baz1a-in-1 | 941521-45-5 | 99.9% | 340.36 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY141890
Encompass_Preferred
BAZ1A-IN-1 is a potent inhibitor of BAZ1A (bromodomain-containing protein) with a KD value of 0.52 μM against the BAZ1A bromodomain. It exhibits good anti-viability activity against cancer cell lines that express high levels of BAZ1A, but shows weak or no activity against cancer cells with low BAZ1A expression.
  • Inhibits BAZ1A bromodomain
  • Shows anti-viability activity against cancer cell lines with high BAZ1A expression
  • Weak or no activity against cancer cells with low BAZ1A expression
  • Provided as a solid
  • High purity
Catalog No. 50-003-64171