Learn More
Cayman Chemical BazedoxIfen-d4 1mg
SDP
Supplier:  Cayman Chemical 225541 MG
Encompass_Preferred
An internal standard for the quantification of bazedoxifene by GC- or LC-MS
Catalog No. 50-298-6533