Learn More
Cayman Chemical BazedoxIfenacetate 10mg
SDP
Supplier:  Cayman Chemical 1500510 MG
Encompass_Preferred
An indole-based SERM that binds to both ERα (IC50 = 26 nM) and ERβ (IC50 = 99 nM); antagonizes 17β-estradiol-dependent and hormone-independent growth of breast cancer cell proliferation (80% reduction with 10 nM) in a manner related to cell cycle arrest and downregulation of cyclin D1 and ERα
Catalog No. 50-297-1889