Learn More
Selleck Chemical LLC Bazedoxifene (WAY-140424) HCl S2128-10mg
SDP
Supplier:  Selleck Chemical LLC S212810MG
Encompass_Preferred
Bazedoxifene HCl (WAY-140424 TSE-424) is a novel non-steroidal indole-based estrogen receptor modulator (SERM) binding to both ER and ER with IC50 of 23 nM and 89 nM respectively
Catalog No. 50-136-2094