Learn More
Apexbio Technology LLC Bazedoxifene HCl 198480-56-7 10mg
SDP
Supplier:  Apexbio Technology LLC B151910
Encompass_Preferred
Bazedoxifene HCl (198480-56-7) is a non-steroidal selective estrogen receptor modulator (SERM) targeting estrogen receptor subtypes ER and ER It is designed to bind ERs thereby modulating estrogen receptor-mediated signaling pathways Bazedoxifene HCl exerts its biological activity primarily by antagonizing ER -mediated transcriptional activity thereby inhibiting estradiol-induced cell proliferation In cell-based studies Bazedoxifene HCl demonstrates inhibition of cell proliferation in human breast cancer (MCF-7) ovarian (CHO) hepatic (HepG2) and neuronal (GT1-7) cell lines The compound exhibits binding affinities (Ki) of 26 nM for ER and 99 nM for ER Based on these pharmacological properties Bazedoxifene HCl holds research potential in osteoporosis and estrogen receptor-mediated diseases including breast ovarian and uterine cancers
Catalog No. 50-000-92367