Learn More
Cayman Chemical BB22 5hydroxyIsoquInolIn I 1mg
SDP
Supplier:  Cayman Chemical 145421 MG
Encompass_Preferred
Differs from BB-22 structurally by having an isoquinoline ring attached at the five carbon instead of a quinoline group attached through the eight position of the ring; intended for forensic and research applications
Catalog No. 50-297-1526