Learn More
Cayman Chemical BB22 6hydroxyIsoquInolIn I 5mg
SDP
Supplier:  Cayman Chemical 145435 MG
Encompass_Preferred
Differs from BB-22 structurally by having the quinoline group replaced with an isoquinoline group attached at its six position; intended for forensic and research applications
Catalog No. 50-297-0133