Learn More
Selleck Chemical LLC BBC0403 E4713-1G
SDP
Supplier:  Selleck Chemical LLC E47131G
Encompass_Preferred
BBC0403 is a small-molecule selective inhibitor of Bromodomain-containing protein 2(BRD2) with Kds of 7 64 M and 41 37 M for BRD2 (BD2) and BRD2 (BD1) respectively It is a potential intra-articular injectable therapeutic agent for treating osteoarthritis (OA)
Catalog No. 50-000-39461