Learn More
Medchemexpress LLC BBT 100mg | 100mg
SDP
Supplier:  Medchemexpress LLC HYQ40876100MG
Encompass_Preferred
BBT is an enhancer of impaired glucose-stimulated insulin secretion (GSIS) BBT exhibits anti-hyperglycemia activity and protects -cells from cytokine- or streptozotocin (STZ (HY-13753))-induced cell death in type 2 diabetes models BBT acts function via cAMP/PKA and long-lasting (L-type) voltage-dependent Ca2 channel/CaMK2 pathway[1]
Catalog No. 50-003-21633